Skip to Content
Merck
All Photos(1)

Key Documents

41570

Sigma-Aldrich

3-(N,N-Dimethyloctadecylammonio)propanesulfonate

≥99.0% (TLC)

Synonym(s):

3-(N,N-Dimethylstearylammonio)propanesulfonate, 3-(Stearyl-dimethylammonio)propanesulfonate, N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, SB3-18, Stearyl sulfobetaine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
419.71
Beilstein:
4152035
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

zwitterionic

Quality Level

Assay

≥99.0% (TLC)

form

powder

mol wt

419.71 g/mol

technique(s)

thin layer chromatography (TLC): suitable

mp

257-260 °C (dec.) (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC[N](C)(C)CCCS(O)(=O)=O

InChI

1S/C23H49NO3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21-24(2,3)22-20-23-28(25,26)27/h4-23H2,1-3H3

InChI key

DIROHOMJLWMERM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(N,N-Dimethyloctadecylammonio)propanesulfonate is a zwitterionic surfactant, useful in membrane solubilization studies. It does not absorb UV light, hence interference with spectrophotometric analysis of proteins can be omitted.

Application

3-(N,N-Dimethyloctadecylammonio)propanesulfonate in combination with bis(2-ethylhexyl) sulfosuccinate sodium salt (AOT) forms an organic mixed micellar phase, which may be used for the extraction of erythromycin from the aqueous phase. The extraction performance and other parameters affecting erythromycin solubilization are evaluated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The solubilization and extraction parameters of erythromycin from aqueous phase into mixed AOT/SB3-18 reverse micelle phase
Mohamad-Aziz NS and Sakinah MMA
Chemical Engineering Research Bulletin, 123-128 (2017)
Disassembling single-walled carbon nanotube bundles by dipole/dipole electrostatic interactions.
Fugetsu B, et al.
Chemistry Letters (Jpn), 34(9), 1218-1219 (2005)
Yan Q Chen et al.
Biochemical and biophysical research communications, 534, 498-503 (2020-11-27)
We previously demonstrated that angiopoietin-like 8 (ANGPTL8) forms a localized complex with ANGPTL4 to reduce its lipoprotein lipase (LPL)-inhibitory activity and enable increased postprandial uptake of fatty acids (FA) into adipose tissue. Because prolonged cold exposure may increase adipose tissue
Solubilization of membrane proteins by sulfobetaines, novel zwitterionic surfactants.
A Gonenne et al.
Analytical biochemistry, 87(1), 28-38 (1978-06-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service