Skip to Content
Merck
All Photos(1)

Key Documents

41566

Sigma-Aldrich

(2R,5R)-(−)-trans-2,5-Dimethylpyrrolidine

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
Beilstein:
4652641
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥97.0% (GC)

optical activity

[α]20/D −19.0±1.5°, c = 2% in methylene chloride

refractive index

n20/D 1.431

density

0.852 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H]1CC[C@@H](C)N1

InChI

1S/C6H13N/c1-5-3-4-6(2)7-5/h5-7H,3-4H2,1-2H3/t5-,6-/m1/s1

InChI key

ZEBFPAXSQXIPNF-PHDIDXHHSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Important chiral auxiliary with a pseudo C2 axis; diastereoselective alkylation of ketone enamines [1]; Highly stereoselective addition reactions of the enamine of β-keto esters (vinylogous urethanes) [2]; Radical reactions of unsaturated amides[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.H. Schlessinger et al.
The Journal of Organic Chemistry, 52, 708-708 (1987)
R.H. Schlessinger et al.
Tetrahedron Letters, 29, 1489-1489 (1988)
N.A. Porter et al.
Journal of the American Chemical Society, 111, 8311-8311 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service