Skip to Content
Merck
All Photos(1)

Key Documents

413674

Sigma-Aldrich

Lanthanum(III) iodide

AnhydroBeads, −10 mesh, 99.9% trace metals basis

Synonym(s):

Lanthanum triiodide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 393.00

CHF 393.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CHF 393.00

About This Item

Linear Formula:
LaI3
CAS Number:
Molecular Weight:
519.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
synthesis grade

CHF 393.00


Check Cart for Availability

Request a Bulk Order

grade

synthesis grade

Quality Level

product line

AnhydroBeads

Assay

99.9% trace metals basis

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤1500.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

mp

772 °C (lit.)

density

5.63 g/mL at 25 °C (lit.)

SMILES string

I[La](I)I

InChI

1S/3HI.La/h3*1H;/q;;;+3/p-3

InChI key

KYKBXWMMXCGRBA-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service