Skip to Content
Merck
All Photos(1)

Key Documents

411477

Sigma-Aldrich

2-Methoxyethyl cyanoacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CO2CH2CH2OCH3
CAS Number:
Molecular Weight:
143.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.434 (lit.)

bp

98-100 °C/1 mmHg (lit.)

density

1.127 g/mL at 25 °C (lit.)

functional group

ester
ether
nitrile

SMILES string

COCCOC(=O)CC#N

InChI

1S/C6H9NO3/c1-9-4-5-10-6(8)2-3-7/h2,4-5H2,1H3

InChI key

SGLKIEOMYXTGBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxyethyl cyanoacetate (3-Oxo-3-(2-methoxyethoxy)propanenitrile) is 2-methoxyethyl ester of 2-cyanoacetic acid.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup - (External MSDS)

Flash Point(C)

113 °C - closed cup - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service