Skip to Content
Merck
All Photos(1)

Key Documents

410616

Sigma-Aldrich

4-Benzyloxy-2(1H)-pyridone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO2
CAS Number:
Molecular Weight:
201.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

201-203 °C (lit.)

SMILES string

O=C1NC=CC(OCc2ccccc2)=C1

InChI

1S/C12H11NO2/c14-12-8-11(6-7-13-12)15-9-10-4-2-1-3-5-10/h1-8H,9H2,(H,13,14)

InChI key

DOVNUEPFPBWTSV-UHFFFAOYSA-N

General description

N-alkylation of 4-benzyloxy-2(1H)-pyridone with the corresponding benzyl halides has been reported.

Application

4-Benzyloxy-2(1H)-pyridone may be used for the synthesis of novel 2-pyridone derivatives. It may be used for the alkylation of the pyridine nitrogen under mild and anhydrous conditions, via reaction with n-Bu4I and KOBu-t.†

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 46, 7917-7917 (2005)
Suresh K Tipparaju et al.
Bioorganic & medicinal chemistry letters, 18(12), 3565-3569 (2008-05-24)
Enoyl-ACP reductase (ENR), the product of the FabI gene, from Bacillus anthracis (BaENR) is responsible for catalyzing the final step of bacterial fatty acid biosynthesis. A number of novel 2-pyridone derivatives were synthesized and shown to be potent inhibitors of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service