Skip to Content
Merck
All Photos(1)

Key Documents

406414

Sigma-Aldrich

Perfluorononane

97%

Synonym(s):

Eicosafluorononane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7CF3
CAS Number:
Molecular Weight:
488.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.276 (lit.)

bp

125-126 °C (lit.)

density

1.799 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C9F20/c10-1(11,2(12,13)4(16,17)6(20,21)8(24,25)26)3(14,15)5(18,19)7(22,23)9(27,28)29

InChI key

UVWPNDVAQBNQBG-UHFFFAOYSA-N

General description

Perfluorononane is a common liquid perfluoro-n-alkane. It is a potent greenhouse gas and environment and health hazards posed by its toxic decomposition products have been evaluated.[1] Perfluorononane is reported as an ideal oral contrast agent since it is biologically inert, immiscible with water, and also has a low viscosity and surface tension.[2] Intramolecular electronic environment of perfluorononane using 13C and 19F NMR techniques in combination with theoretical calculations of molecular orbitals has been reported.[3]

Application

Perfluorononane may be employed in the interface between active and buffer layer to improve the performance of polymer photovoltaic cells.[4] It may be employed as selective and specific markers for monitoring the gastric emptying and gastrointestinal (GI) transit times in mice and humans by using 27Al and 19F nuclear magnetic resonance (NMR).[5]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Roman Schwarz et al.
Magnetic resonance in medicine, 48(2), 255-261 (2002-09-05)
Gastric emptying and gastrointestinal (GI) transit times in mice and humans were monitored noninvasively by using 27Al and 19F nuclear magnetic resonance (NMR). Al(3+) bound to ion-exchange resin and perfluorononane were administered orally as selective and specific markers for the
Chi-Ang Tseng et al.
ACS applied materials & interfaces, 7(12), 6683-6689 (2015-03-15)
In this study, we investigate the effects of fluorinated poly(3,4-ethylene dioxythiophene):poly(styrenesulfonate) buffer layer on the performance of polymer photovoltaic cells. We demonstrate for the first time, the deterioration of the device performance can be effectively mended by modifying the interface
Roman Schwarz et al.
Magnetic resonance in chemistry : MRC, 42(6), 512-517 (2004-05-12)
Liquid perfluorocarbons exhibit unique physical-chemical characteristics such as extraordinary stability, combined hydrophobia and lipophobia, low surface tension and a capacity to carry large quantities of gas. They have found widespread use in industry, medicine and biology even though the molecular
Fernando Soto et al.
Small (Weinheim an der Bergstrasse, Germany), 14(49), e1803266-e1803266 (2018-10-29)
Current technologies for managing acute and chronic pain have focused on reducing the time required for achieving high therapeutic efficiency. Herein a wearable transdermal patch is introduced, employing an acoustic droplet vaporization (ADV) methodology, as an effective noninvasive transdermal platform
Fernando Soto et al.
ACS nano, 10(1), 1522-1528 (2015-12-23)
Acoustically triggered microcannons, capable of loading and firing nanobullets (Nbs), are presented as powerful microballistic tools. Hollow conically shaped microcannon structures have been synthesized electrochemically and fully loaded with nanobullets made of silica or fluorescent microspheres, and perfluorocarbon emulsions, embedded

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service