Skip to Content
Merck
All Photos(1)

Key Documents

406198

Sigma-Aldrich

1,4-Butanediol vinyl ether

contains 0.01% potassium hydroxide as stabilizer, 99%

Synonym(s):

1,4-Butanediol monovinyl ether, 4-(Ethenyloxy)-1-butanol, 4-(Vinyloxy)-1-butanol, 4-Hydroxybutyl vinyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHO(CH2)4OH
CAS Number:
Molecular Weight:
116.16
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

contains

0.01% potassium hydroxide as stabilizer

refractive index

n20/D 1.444 (lit.)

bp

95 °C/20 mmHg (lit.)

density

0.939 g/mL at 25 °C (lit.)

SMILES string

OCCCCOC=C

InChI

1S/C6H12O2/c1-2-8-6-4-3-5-7/h2,7H,1,3-6H2

InChI key

HMBNQNDUEFFFNZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service