Skip to Content
Merck
All Photos(1)

Key Documents

404713

Sigma-Aldrich

Chlorodiisobutyloctadecylsilane

technical grade, 85%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CHF 88.10

CHF 88.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
CHF 88.10

About This Item

Linear Formula:
CH3(CH2)17Si[CH2CH(CH3)2]2Cl
CAS Number:
Molecular Weight:
431.25
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

CHF 88.10


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

85%

form

liquid

refractive index

n20/D 1.459 (lit.)

bp

140-170 °C/0.05 mmHg (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC[Si](Cl)(CC(C)C)CC(C)C

InChI

1S/C26H55ClSi/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-28(27,23-25(2)3)24-26(4)5/h25-26H,6-24H2,1-5H3

InChI key

LAMXXRLSKVGVCO-UHFFFAOYSA-N

Other Notes

remainder octadecane

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service