Skip to Content
Merck
All Photos(1)

Documents

400165

Sigma-Aldrich

Dichloro(1,10-phenanthroline)platinum(II)

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl2N2Pt
CAS Number:
Molecular Weight:
446.19
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

reaction suitability

core: platinum
reagent type: catalyst

SMILES string

Cl[Pt]Cl.c1cnc2c(c1)ccc3cccnc23

InChI

1S/C12H8N2.2ClH.Pt/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;;/h1-8H;2*1H;/q;;;+2/p-2

InChI key

VONLGUDYJIAGQI-UHFFFAOYSA-L

Application

Reactant for:
  • Mixed-metal complexes incorporating platinum and lanthanide centers
  • Synthesis of platinum complexes containing a chelating di-fluoro-substituted thiourea ligand
  • Adduct formation between ternary Pt(III)-amino acid- aromatic diimine complexes and flavin mononucleotide

Catalyst for the selective synthesis of monoorganotin trihalides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service