Skip to Content
Merck
All Photos(1)

Key Documents

399949

Sigma-Aldrich

Vanadium(V) oxyfluoride

99%

Synonym(s):

Vanadium(V) oxytrifluoride, Vanadyl trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
VOF3
CAS Number:
Molecular Weight:
123.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

powder

reaction suitability

reagent type: catalyst
core: vanadium

solubility

H2O: insoluble(lit.)

application(s)

battery manufacturing

SMILES string

F[V](F)(F)=O

InChI

1S/3FH.O.V/h3*1H;;/q;;;;+3/p-3

InChI key

FXGIDXZUDLFPLD-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micah J Niphakis et al.
Organic letters, 13(2), 196-199 (2010-12-15)
A highly convergent strategy to prepare phenanthroindolizidines is reported involving three consecutive C-C coupling reactions. This sequence features a novel VOF(3)-mediated aryl-alkene coupling in the final step, which enables regioselective preparation of C5-substituted phenanthroindolizidines for the first time. This strategy
M D Zidan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56(14), 2693-2698 (2001-01-06)
A new route has been devised, leading to the production of VOX3 molecules where X=F, Br and I by an on-line process using vanadium oxytrichloride, VOCl3 as a starting compound passed over the following heated salts NaF, KBr and KI

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service