Skip to Content
Merck
All Photos(2)

Key Documents

399868

Sigma-Aldrich

Hafnium(IV) fluoride

99.9% trace metals basis (purity excludes up to ~1% zirconium)

Synonym(s):

Hafnium tetrafluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HfF4
CAS Number:
Molecular Weight:
254.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
powder

Assay

99.9% trace metals basis (purity excludes up to ~1% zirconium)

form

powder

impurities

≤2000.0 ppm Trace Metal Analysis (excluding Zr content)

SMILES string

F[Hf](F)(F)F

InChI

1S/4FH.Hf/h4*1H;/q;;;;+4/p-4

InChI key

QHEDSQMUHIMDOL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service