389005
Butoxycarbonylmethyl butyl phthalate
technical grade
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
2-[CH3(CH2)3O2CCH2O2C]C6H4CO2(CH2)3CH3
CAS Number:
Molecular Weight:
336.38
EC Number:
MDL number:
UNSPSC Code:
12162002
grade
technical grade
vapor density
11.8 (vs air)
vapor pressure
3.9 mmHg ( 200 °C)
refractive index
n20/D 1.49 (lit.)
bp
219 °C/5 mmHg (lit.)
density
1.1 g/mL at 25 °C (lit.)
SMILES string
CCCCOC(=O)COC(=O)c1ccccc1C(=O)OCCCC
Application
Useful poly(vinyl chloride) plasticizer
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service