Skip to Content
Merck
All Photos(1)

Key Documents

388823

Sigma-Aldrich

3-(N,N-Dimethylmyristylammonio)propanesulfonate

97%

Synonym(s):

3-(N,N-Dimethyltetradecylammonio)propanesulfonate, 3-(Myristyldimethylammonio)propanesulfonate, N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, Myristyl sulfobetaine, SB3-14

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
363.60
Beilstein:
6419727
EC Number:
MDL number:
UNSPSC Code:
12352103

description

zwitterionic

Assay

97%

mol wt

micellar avg mol wt 30,200

aggregation number

83

CMC

0.1-0.4 mM (20-25°C)

SMILES string

CCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C19H41NO3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-17-20(2,3)18-16-19-24(21,22)23/h4-19H2,1-3H3

InChI key

BHATUINFZWUDIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zwitterionic detergent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Russell-Harde et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 15(1), 31-37 (1995-01-01)
A new method for purifying human interferon-beta SER17 from E. coli-derived inclusion bodies has been developed. This procedure eliminates the need for strong denaturants, such as sodium dodecyl sulfate or chaotropes. The procedure makes use of a nondenaturing detergent and
L Goetsch et al.
Vaccine, 19(28-29), 4036-4042 (2001-06-28)
Respiratory syncytial virus (RSV) is a major respiratory pathogen responsible for severe pulmonary disease. We have developed a parenterally administered vaccine, BBG2Na, which is currently in a phase III clinical trial. BBG2Na comprises residues 130--230 of RSV-A G protein (G2Na)
S M Halling et al.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 13(5), 408-412 (2001-10-03)
Serologic responses to the newly introduced rough Brucella abortus vaccine strain RB51 have been determined in a dot-blot format using gamma-irradiated RB51 cells as the antigen. Because gamma-irradiated cells are not easily prepared and the signal from cells was not
J T Poolman et al.
Antonie van Leeuwenhoek, 53(6), 413-419 (1987-01-01)
Outer membrane proteins (OMPs) were isolated from meningococcal strain H44/76 (B:15:P1.16) by detergent extraction of bacteria. A final product containing class 1 (P1.16), 3(15), 4 OMPs and 5% (w/w) lipooligosaccharide was obtained. Two experimental vaccines were prepared: OMP-detergent and OMP-C
G J Speijers et al.
Vaccine, 7(4), 364-368 (1989-08-01)
In order to assess the systemic toxicity of Zwittergent 3-14, a detergent used in the protein-detergent complex vaccines for its attractive immunogenic properties, a subacute toxicity study was performed. In this 4-week toxicity experiment five female and five male rats

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service