Skip to Content
Merck
All Photos(1)

Documents

379131

Sigma-Aldrich

Bis(cyclopentadienyl)magnesium(II)

purified by sublimation, 97%

Synonym(s):

Bis(cyclopentadienyl)magnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(C5H5)2
CAS Number:
Molecular Weight:
154.49
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

purified by

sublimation

reaction suitability

core: magnesium

mp

180 °C (dec.) (lit.)

SMILES string

[Mg].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.Mg/c2*1-2-4-5-3-1;/h2*1-5H;

InChI key

USZGMDQWECZTIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Sol. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service