Skip to Content
Merck
All Photos(1)

Key Documents

377082

Sigma-Aldrich

p-Xylylenebis(tetrahydrothiophenium chloride)

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24Cl2S2
CAS Number:
Molecular Weight:
351.40
MDL number:
UNSPSC Code:
12162002

grade

technical grade

Assay

90%

mp

157-161 °C (lit.)

SMILES string

[Cl-].[Cl-].C1CC[S+](C1)Cc2ccc(C[S+]3CCCC3)cc2

Looking for similar products? Visit Product Comparison Guide

Application

Poly(p-phenylene vinylene) precursor.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 25, 38-38 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service