Skip to Content
Merck
All Photos(1)

Key Documents

375152

Sigma-Aldrich

Benzeneselenol

97%

Synonym(s):

Phenyl selenol, Phenyl selenomercaptan, Selenophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SeH
CAS Number:
Molecular Weight:
157.07
Beilstein:
385715
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.616 (lit.)

bp

71-72 °C/18 mmHg (lit.)

density

1.479 g/mL at 25 °C (lit.)

SMILES string

[SeH]c1ccccc1

InChI

1S/C6H6Se/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

WDODWFPDZYSKIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzeneselenol is an aryl selenol. Diphenyl diselenide-promoted radical addition of benzeneselenol to inactivated acetylenes upon irradiation through Pyrex with a tungsten lamp has been reported. Catalysis of stannane-mediated radical chain reactions by benzeneselenol, generated in situ by reduction of diphenyl diselenide with tributyltin hydride, has been reported.

Application

Benzeneselenol may be used as starting reagent in the synthesis of monoseleno-substituted 1,3-dienes. It may be used in the synthesis of cationic chalcogenolato-bridged diruthenium complexes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin P Johnpeter et al.
Inorganic chemistry, 52(23), 13663-13673 (2013-11-20)
A series of cationic chalcogenolato-bridged diruthenium complexes [(η(6)-p-MeC6H4Pr(i))2Ru2(μ-EC6H5)3](+) (E = S, 1; E = Se, 2; E = Te, 3) has been obtained in ethanol from the reaction of (η(6)-p-MeC6H4Pr(i))2Ru2(μ-Cl)2Cl2 with benzenethiol, benzeneselenol, and sodium tellurophenolate, respectively. The thiolato and
Nikolay V Orlov et al.
The Journal of organic chemistry, 79(24), 12111-12121 (2014-10-08)
A unique Ni-catalyzed transformation is reported for the one-pot highly selective synthesis of previously unknown monoseleno-substituted 1,3-dienes starting from easily available terminal alkynes and benzeneselenol. The combination of a readily available catalyst precursor, Ni(acac)2, and an appropriately tuned phosphine ligand
Brittany Trang et al.
Journal of the American Chemical Society, 140(42), 13892-13903 (2018-09-29)
Silver metal exposed to the atmosphere corrodes and becomes tarnished as a result of oxidation and precipitation of the metal as an insoluble salt. Tarnish has so poor a reputation that the word itself connotes corruption and disrespectability; however, tarnishing
Lorenzo Maserati et al.
Nanoscale, 13(1), 233-241 (2020-12-18)
Hybrid quantum wells are electronic structures where charge carriers are confined along stacked inorganic planes, separated by insulating organic moieties. 2D quantum-confined hybrid materials are of great interest from a solid-state physics standpoint because of the rich many-body phenomena they
Diphenyl diselenide-promoted radical addition of benzeneselenol to acetylenes.
Ogawa A, et al.
Tetrahedron Letters, 33(10), 1329-1332 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service