Skip to Content
Merck
All Photos(1)

Key Documents

370053

Sigma-Aldrich

4-Isothiocyanatophenyl 4-pentylbicyclo[2.2.2]octane-1-carboxylate

liquid crystal (nematic), 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO2S
CAS Number:
Molecular Weight:
357.51
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

form

liquid crystal (nematic)

mp

73-113.5 °C (lit.)

transition temp

nematic phase to isotropic phase 113.5 °C
crystalline phase to nematic phase 74.5 °C

SMILES string

CCCCC[C@@]12CC[C@@](CC1)(CC2)C(=O)Oc3ccc(cc3)N=C=S

InChI

1S/C21H27NO2S/c1-2-3-4-9-20-10-13-21(14-11-20,15-12-20)19(23)24-18-7-5-17(6-8-18)22-16-25/h5-8H,2-4,9-15H2,1H3/t20-,21-

InChI key

BXZVNXOHCXCVAG-MEMLXQNLSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chiral nematic liquid crystal (N*-LC) facilitates asymmetric acetylene polymerization, yielding helical chains and fibrils in polyacetylene film.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service