Skip to Content
Merck
All Photos(1)

Key Documents

366684

Sigma-Aldrich

Dimethyl trans-cyclohexane-1,4-dicarboxylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CO2CH3)2
CAS Number:
Molecular Weight:
200.23
Beilstein:
2213375
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

67-69 °C (lit.)

SMILES string

COC(=O)[C@@H]1CC[C@H](CC1)C(=O)OC

InChI

1S/C10H16O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h7-8H,3-6H2,1-2H3/t7-,8-

InChI key

LNGAGQAGYITKCW-ZKCHVHJHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl trans-cyclohexane-1,4-dicarboxylate is an unsaturated ester. Sterically favored conformations of the dimethyl trans-cyclohexane-1,4-dicarboxylate molecule has been reported.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RIS Models and Unperturbed Chain Properties of Polyesters Containing 1, 3-Cyclobutylene and 1, 4-Cyclohexylene Linkages.
Sulatha MS and Natarajan U.
Macromolecular Theory and Simulations, 12(1), 61-71 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service