Skip to Content
Merck
All Photos(2)

Documents

362204

Sigma-Aldrich

Dichloro(1,10-phenanthroline)copper(II)

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl2CuN2
CAS Number:
Molecular Weight:
314.66
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

core: copper
reagent type: catalyst

SMILES string

Cl[Cu]Cl.c1cnc2c(c1)ccc3cccnc23

InChI

1S/C12H8N2.2ClH.Cu/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;;/h1-8H;2*1H;/q;;;+2/p-2

InChI key

PHECBGBJCLDCMF-UHFFFAOYSA-L

Application

Catalyst for:
  • Oxidative carbonylation of methanol
  • Cyclohexane and toluene oxidation
  • Selective oxidation of tetralin

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thor C Møller et al.
Scientific reports, 8(1), 10414-10414 (2018-07-12)
G protein coupled receptors (GPCRs) play essential roles in intercellular communication. Although reported two decades ago, the assembly of GPCRs into dimer and larger oligomers in their native environment is still a matter of intense debate. Here, using number and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service