Skip to Content
Merck
All Photos(1)

Documents

359955

Sigma-Aldrich

Boron trifluoride acetic acid complex

98%

Synonym(s):

Boron trifluoride-acetic acid (1:2)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · 2CH3COOH
CAS Number:
Molecular Weight:
187.91
Beilstein:
3686517
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

reaction suitability

core: boron
reagent type: catalyst

refractive index

n20/D 1.368 (lit.)

density

1.353 g/mL at 25 °C (lit.)

SMILES string

CC(O)=O.CC(O)=O.FB(F)F

InChI

1S/2C2H4O2.BF3/c2*1-2(3)4;2-1(3)4/h2*1H3,(H,3,4);

InChI key

COTMJGCQSLZICX-UHFFFAOYSA-N

Related Categories

General description

Boron trifluoride acetic acid is a versatile Lewis acid catalyst in organic synthesis with various applications. It is used as a catalyst in various reactions such as esterification, Friedel-Crafts reactions, oxidation, Fries rearrangement, and polymerization reactions.

Application

Boron trifluoride acetic acid complex is used as a Lewis acid catalyst in the Fries rearrangement reaction, which is a critical step in the total synthesis of furanaphin. It is also used as a co-catalyst in the intramolecular amination reaction of primary sulfonamides to form cyclic carbamates.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service