Skip to Content
Merck
All Photos(1)

Documents

357499

Sigma-Aldrich

Tin(IV) chloride solution

1.0 M in heptane

Synonym(s):

Stannic chloride, Tin tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl4
CAS Number:
Molecular Weight:
260.52
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

core: tin
reagent type: Lewis acid
reagent type: catalyst

concentration

1.0 M in heptane

density

0.874 g/mL at 25 °C

SMILES string

Cl[Sn](Cl)(Cl)Cl

InChI

1S/4ClH.Sn/h4*1H;/q;;;;+4/p-4

InChI key

HPGGPRDJHPYFRM-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuyuki Kita et al.
The Journal of organic chemistry, 71(14), 5191-5197 (2006-07-01)
The Lewis acid-promoted rearrangement of 2,2,3,3-tetrasubstituted 2,3-epoxy alcohols with several kinds of protecting groups was investigated. When SnCl4 is used as a Lewis acid, the reaction proceeds in a regio- and stereo-controlled manner to afford two types of carbonyl compounds
L Malaguarnera et al.
Journal of cellular biochemistry, 93(1), 197-206 (2004-09-08)
The pituitary hormone prolactin (PRL) is a multifunctional polypeptide which exerts a role on cell proliferation and may also contribute to cell differentiation. PRL is also produced by immune cells and is regarded as a key component of the neuroendocrine-immune
Gerry A Griffith et al.
Journal of the American Chemical Society, 128(40), 13130-13141 (2006-10-05)
Difluorinated alkenoate ethyl 3,3-difluoro-2-(N,N-diethylcarbamoyloxy)-2-propenoate reacts rapidly and in high yield with furan and a range of substituted furans in the presence of a tin(IV) catalyst. Non-fluorinated congener 2-(N,N-diethylcarbamoyloxy)-2-propenoate fails to react at all under the same conditions. These reactions have
Gary B Evans et al.
The Journal of organic chemistry, 69(6), 2217-2220 (2004-04-03)
Promotion by Lewis acid of the addition of some aryllithiums to a carbohydrate-based imine, which has allowed a more facile synthesis of some imino-C-nucleoside analogues, is described. Use of the corresponding nitrone does not assist in some cases, but lithiated
Colin O'Brien et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(3), 902-909 (2006-11-07)
The reaction of silylated nucleophiles with 6,1-anhydroglucopyranuronic acid (glucuronic acid 6,1-lactones) catalysed by tin(IV) chloride provides 1,2-trans or 1,2-cis (deoxy)glycosides in a manner dependent on the donor structure. The alpha-glycoside was obtained for reactions of the donor with the 2-acyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service