Skip to Content
Merck
All Photos(1)

Key Documents

349992

Sigma-Aldrich

2-Aminobenzyl cyanide

97%

Synonym(s):

2-Aminophenylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH2CN
CAS Number:
Molecular Weight:
132.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

69-72 °C (lit.)

SMILES string

Nc1ccccc1CC#N

InChI

1S/C8H8N2/c9-6-5-7-3-1-2-4-8(7)10/h1-4H,5,10H2

InChI key

LMDPYYUISNUGGT-UHFFFAOYSA-N

Application

2-Aminobenzyl cyanide may be used in the preparation of oxindole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The chemistry of oxindole.
W C SUMPTER
Chemical reviews, 37, 443-479 (1945-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service