Skip to Content
Merck
All Photos(2)

Key Documents

342939

Sigma-Aldrich

Barium titanate(IV)

pieces, 3-12 mm, 99.8% trace metals basis

Synonym(s):

Barium metatitanate, Titanium barium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaTiO3
CAS Number:
Molecular Weight:
233.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

99.8% trace metals basis

form

pieces
sintered

reaction suitability

core: barium
reagent type: catalyst
reagent type: catalyst
core: titanium

particle size

3-12 mm

density

6.08 g/mL at 25 °C (lit.)

SMILES string

[Ba++].[O-][Ti]([O-])=O

InChI

1S/Ba.3O.Ti/q+2;;2*-1;

InChI key

WNKMTAQXMLAYHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for:
  • Synthesis of high-dielectric polymer composite materials
  • Inverse electron demand Diels-Alder reactions

Catalyst for:
  • Selectivity of methane oxidation
  • Vapor phase intramolecular dehydration

Used for:
  • Synthesis of flexible transparent conductive coatings
  • X7R multilayer ceramic capacitors
Suitable for vacuum deposition.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Layani, M.; Magdassi, S.
Journal of Materials Chemistry, 21, 15378-15378 (2011)
H. Lee, et al.,
Journal of the American Ceramic Society. American Ceramic Society, 94, 1556-1562 (2011)
Y. Shimasaki, et al.,
J. Mol. Catal. A: Chem., 239, 125-129 (2005)
Peter Barber et al.
ACS applied materials & interfaces, 2(9), 2553-2559 (2010-09-02)
We report the preparation of new polymer composite dielectric materials for energy storage applications. New layered 1:1 mixed A+2/Ti4+ metal phenylphosphonates, ATi(O3PC6H5)3, A=Mg, Ca, Sr, Ba, and Pb, have been prepared via a melt route, in which mixed metal oxides
O. A. Timonova and Y. S. Mardeshev,
Russ. J. Gen. Chem., 77, 2214-2214 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service