Skip to Content
Merck
All Photos(1)

Key Documents

341398

Sigma-Aldrich

Lithium-7Li hydroxide monohydrate

97 atom % 7Li

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
7LiOH·H2O
CAS Number:
Molecular Weight:
42.04
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

isotopic purity

97 atom % 7Li

Assay

≥99.9% (titration)

form

solid

density

1.51 g/mL at 25 °C (lit.)

mass shift

depleted

SMILES string

[7Li+].O.[OH-]

InChI

1S/Li.2H2O/h;2*1H2/q+1;;/p-1/i1+0;;

InChI key

GLXDVVHUTZTUQK-GLJCYROLSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Sivakumar et al.
Ultrasonics sonochemistry, 26, 332-339 (2015-03-10)
LiNi0.5Mn1.5O4 was synthesized as a cathode material for Li-ion batteries by a sonochemical reaction followed by annealing, and was characterized by XRD, SEM, HRTEM and Raman spectroscopy in conjunction with electrochemical measurements. Two samples were prepared by a sonochemical process
Ludmila Müllerová et al.
Journal of chromatography. A, 1384, 147-154 (2015-02-11)
Interactions among analyte forms that undergo simultaneous dissociation/protonation and complexation with multiple selectors take the shape of a highly interconnected multi-equilibrium scheme. This makes it difficult to express the effective mobility of the analyte in these systems, which are often
Sergej Repp et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 152, 637-644 (2015-02-25)
ZnO nanoparticles were synthesized by solid state and hydrolysis methods based on the conventional precipitation. In situ growth of ZnO nanoparticles were monitored by photoluminescence spectroscopy (PL). By the help of electron paramagnetic resonance (EPR) technique, detailed analysis of intrinsic
Martina Riesová et al.
Journal of chromatography. A, 1364, 276-288 (2014-09-13)
In this paper we determine acid dissociation constants, limiting ionic mobilities, complexation constants with β-cyclodextrin or heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin, and mobilities of resulting complexes of profens, using capillary zone electrophoresis and affinity capillary electrophoresis. Complexation parameters are determined for both neutral and
Derek Power et al.
Virology, 481, 142-150 (2015-03-18)
IFI44 is an interferon-alfa inducible protein, and is associated with infection of several viruses. However, IFI44 elicits minimal antiviral effects on these viruses, and its exact role is still unknown. Here we show that IFI44 inhibits HIV-1 replication in vitro.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service