Skip to Content
Merck
All Photos(1)

Key Documents

340545

Sigma-Aldrich

1,2,3-Benzotriazin-4(3H)-one

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O
CAS Number:
Molecular Weight:
147.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

216-218 °C (lit.)

SMILES string

O=C1NN=Nc2ccccc12

InChI

1S/C7H5N3O/c11-7-5-3-1-2-4-6(5)8-10-9-7/h1-4H,(H,8,9,11)

InChI key

DMSSTTLDFWKBSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,3-Benzotriazin-4(3H)-one is an 1,2,3-benzotriazine derivative. 1,2,3-Benzotriazin-4(3H)-one undergoes thermal condensation with α-amino acids to yield 3H-1,4-benzodiazepin-(1H,4H)-2,5-diones. 1,2,3-benzotriazin-4(3H)-one on thermolysis yields quinazolino[3,2-c][1,2,3]benzotriazin-8-one.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel one step approach to the synthesis of 3H-1, 4-benzodiazepin-(1H, 4H)-2, 5-diones from 1, 2, 3-benzotriazin-4-(3H)-one.
Gupta R, et al.
Heterocyclic Communications, 9(4), 363-366 (2003)
Thermolysis of 1, 2, 3-benzotriazin-4 (3 H)-one.
Murray AR and Vaughan K.
J. Chem. Soc. Sect. C, 15, 2070-2074 (1970)
E D Woodland et al.
The Journal of pharmacy and pharmacology, 48(2), 223-227 (1996-02-01)
1,2,3-Benzotriazin-4-one derivatives are extensively used as herbicides, insecticides and nematicides. Incorporation of these compounds into the human food chain is a cause for concern, since their toxicity to man is well documented. This work describes the development of a data-base
Angela Corvino et al.
Current medicinal chemistry, 25(27), 3214-3227 (2018-02-10)
The 5-HT1A receptor is a pharmacologically well characterized serotonin receptor subtype and it has long been investigated because of its involvement in several physiopathological mechanisms and treatment of neurological diseases like ansia and depression. Serotonin (5-HT) also shows many non-neural
Margarida S Miranda et al.
The journal of physical chemistry. B, 115(20), 6616-6622 (2011-05-06)
This paper reports an experimental and computational study on the energetics of 1,2,3-benzotriazin-4(3H)-one. The standard (p° = 0.1 MPa) molar enthalpy of formation of solid 1,2,3-benzotriazin-4(3H)-one, at T = 298.15 K, was derived from its standard massic energy of combustion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service