Skip to Content
Merck
All Photos(2)

Key Documents

339407

Sigma-Aldrich

Ethyl 2-nitropropionate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NO2)CO2C2H5
CAS Number:
Molecular Weight:
147.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.421 (lit.)

bp

75-76 °C/9 mmHg (lit.)

density

1.13 g/mL at 25 °C (lit.)

functional group

amine
ester
nitro

SMILES string

CCOC(=O)C(C)[N+]([O-])=O

InChI

1S/C5H9NO4/c1-3-10-5(7)4(2)6(8)9/h4H,3H2,1-2H3

InChI key

ZXBGJDZWJJFFQY-UHFFFAOYSA-N

General description

Ethyl 2-nitropropionate is a nitro ester.[1]

Application

Ethyl 2-nitropropionate was used in the preparation of:
  • (−)-trans-2,5-dimethylprolineethyl ester via enantioselective Michael reaction methyl vinyl ketone using modified dihydroquinine as catalyst[2]
  • novel nitrogen-15-labeled nitrone spin trap, 5-ethoxycarbonyl-5-methyl-1-pyrroline N-oxide ([15N]EMPO) for detecting superoxide anion[3]
  • 2-ethoxycarbonyl-2-nitropentanal[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Zhang et al.
FEBS letters, 473(1), 58-62 (2000-05-10)
We describe the synthesis and biological applications of a novel nitrogen-15-labeled nitrone spin trap, 5-ethoxycarbonyl-5-methyl-1-pyrroline N-oxide ([(15)N]EMPO) for detecting superoxide anion. Superoxide anion generated in xanthine/xanthine oxidase (100 nM min(-1)) and NADPH/calcium-calmodulin/nitric oxide synthase systems was readily detected using EMPO
5-Carboxy-5-methyl-1-pyrroline N-oxide: a spin trap for the hydroxyl radical.
Tsai P, et al.
J. Chem. Soc. Perkin Trans. II, 6, 875-880 (2001)
Jeremy R Duvall et al.
The Journal of organic chemistry, 71(22), 8579-8590 (2006-10-27)
The proposed structures of jenamidines A, B, and C (1-3) were revised to jenamidines A1/A2, B1/B2, and C (8-10). Jenamidines A1/A2 (8) were synthesized from activated proline derivative 43 by conversion to 26 in two steps and 50% overall yield.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service