Skip to Content
Merck
All Photos(1)

Key Documents

338710

Sigma-Aldrich

(−)-1-(9-Fluorenyl)ethyl chloroformate solution

18 mM in acetone, for chiral derivatization

Synonym(s):

(−)-FLEC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClO2
CAS Number:
Molecular Weight:
272.73
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

concentration

18 mM in acetone

density

0.79 g/mL at 25 °C

functional group

chloro

storage temp.

2-8°C

SMILES string

CC(OC(Cl)=O)C1c2ccccc2-c3ccccc13

InChI

1S/C16H13ClO2/c1-10(19-16(17)18)15-13-8-4-2-6-11(13)12-7-3-5-9-14(12)15/h2-10,15H,1H3

InChI key

SFRVOKMRHPQYGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-1-(9-Fluorenyl)ethyl chloroformate is mostly used as an in-capillary derivatization agent for the separation of amino acid (AA) derivatives by micellar electrokinetic chromatography (MEKC).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17.0 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

In-capillary derivatization with (-)-1-(9-fluorenyl) ethyl chloroformate as chiral labeling agent for the electrophoretic separation of amino acids.
Fradi I, et al.
Journal of Chromatography A, 1323, 338-347 (2014)
Radu-Cristian Moldovan et al.
Journal of chromatography. A, 1513, 1-17 (2017-08-02)
Over the last 30years, (±)-1-(9-fluorenyl)ethyl chloroformate ((±)-FLEC) was used as a chiral derivatizing agent in various analytical applications involving a wide range of endogenous, pharmaceutical and environmentally relevant molecules. This comprehensive review aims to present all the significant aspects related
Radu-Cristian Moldovan et al.
Journal of chromatography. A, 1564, 199-206 (2018-06-19)
A targeted CE-MS approach was developed for the chiral analysis of biologically relevant amino acids in artificial cerebrospinal fluid (aCSF). In order to achieve chiral resolution, the five amino acids (Ser, Asn, Asp, Gln and Glu) were derivatized with (+)-1-(9-fluorenyl)ethyl
Radu-Cristian Moldovan et al.
Journal of chromatography. A, 1590, 80-87 (2019-01-15)
D-amino acids (AA) analysis is becoming more and more relevant for metabolomics, therefore new analytical tools need to be developed. A common approach to achieve AA enantioseparation is chiral derivatization. Among the chiral derivatization reagents, (+) or (-)-1-(9-fluorenyl) ethyl chloroformate
Radu-Cristian Moldovan et al.
Journal of chromatography. A, 1467, 400-408 (2016-08-25)
In the context of bioanalytical method development, process automatization is nowadays a necessity in order to save time, improve method reliability and reduce costs. For the first time, a fully automatized micellar electrokinetic chromatography-mass spectrometry (MEKC-MS) method with in-capillary derivatization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service