Skip to Content
Merck
All Photos(1)

Key Documents

335177

Sigma-Aldrich

2-Methoxyphenyl acetate

98%

Synonym(s):

O-Acetylguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4OCH3
CAS Number:
Molecular Weight:
166.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.514 (lit.)

bp

122-124 °C/18 mmHg (lit.)

density

1.129 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1OC(C)=O

InChI

1S/C9H10O3/c1-7(10)12-9-6-4-3-5-8(9)11-2/h3-6H,1-2H3

InChI key

BHJHPYFAYGAPLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methoxyphenyl acetate may be used in chemical synthesis.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service