Skip to Content
Merck
All Photos(1)

Key Documents

334111

Sigma-Aldrich

Dipentene

technical, for use as solvent (for the paint industry), mixture of various terpenes

Synonym(s):

(±)-Limonene, p-Mentha-1,8-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
3587825
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

for use as solvent (for the paint industry)
technical

vapor density

>4.7 (80 °C, vs air)

vapor pressure

1 mmHg ( 20 °C)

form

liquid

autoignition temp.

458 °F

composition

Major isomer: 3-Carene
Minor isomers: Limonene, β-pinene and α-pinene

expl. lim.

0.7-6.1 %, 150 °F

refractive index

n20/D 1.473 (lit.)

bp

170-180 °C (lit.)

density

0.86 g/mL at 20 °C (lit.)

SMILES string

CC(=C)C1CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3

InChI key

XMGQYMWWDOXHJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dipentene (Limonene) is a monoterpene olefin having potential applications in polymer and fuel chemistry. It is also widely used as flavoring and fragrance agent. Dipentene (limonene) is a promising green solvent. Kinetics of the reactions of limonene with OH and OD radicals has been investigated in a low pressure flow tube reactor coupled with a quadrupole mass spectrometer.

Application

Dipentene was employed as a solvent in the reaction media for enzymatic synthesis of phosphatidylserine.

Other Notes

Contains a mixture of terpenes

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tristan Braure et al.
The journal of physical chemistry. A, 118(40), 9482-9490 (2014-09-12)
The kinetics of the reactions of limonene with OH and OD radicals has been studied using a low-pressure flow tube reactor coupled with a quadrupole mass spectrometer: OH + C10H16 → products (1), OD + C10H16 → products (2). The
Yan-Hong Bi et al.
Biotechnology letters, 37(1), 115-119 (2014-09-13)
The bio-based solvents limonene and p-cymene obtained from citrus waste were innovatively employed as the reaction media for enzymatic synthesis of phosphatidylserine. (R)-(+)-Limonene, which is available in large quantities from citrus waste, and its close derivative p-cymene, are promising green
Patrizia A d'Alessio et al.
Life sciences, 92(24-26), 1151-1156 (2013-05-15)
To further explore the anti-inflammatory properties of d-Limonene. A rat model was used to compare evolution of TNBS (2,5,6-trinitrobenzene sulfonic acid)-induced colitis after oral feeding with d-Limonene compared to ibuprofen. Peripheral levels of TNF-α (Tumor Necrosis Factor alpha) were assessed
Young Woo Kim et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 16(1), 17-38 (2013-04-12)
d-Limonene, a major constituent of citrus oils, is a monoterpene widely used as a flavor/fragrance additive in cosmetics, foods, and industrial solvents as it possesses a pleasant lemon-like odor. d-Limonene has been designated as a chemical with low toxicity based
Johanna Bråred Christensson et al.
Contact dermatitis, 68(4), 214-223 (2013-03-21)
Limonene is a common fragrance terpene that, in its pure form, is not allergenic or is a very weak allergen. However, limonene autoxidizes on air exposure, and the oxidation products can cause contact allergy. Oxidized R-limonene has previously been patch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service