Skip to Content
Merck
All Photos(1)

Documents

328812

Sigma-Aldrich

Acetoacetamide

97%

Synonym(s):

α-Acetylacetamide, 3-Oxobutanamide, 3-Oxobutyramide, Acetoacetic acid amide, Acetylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CONH2
CAS Number:
Molecular Weight:
101.10
Beilstein:
1560550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

53-56 °C (lit.)

solubility

water: soluble 1 g/10 mL, clear, colorless

SMILES string

CC(=O)CC(N)=O

InChI

1S/C4H7NO2/c1-3(6)2-4(5)7/h2H2,1H3,(H2,5,7)

InChI key

GCPWJFKTWGFEHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetoacetamide is a degradation product of acesulfame-K, sweetener. Acetoacetamide undergoes alkaline hydrolysis to form salt of acetoacetic acid.

Application

Acetoacetamide was used in a study to assess the stability of artificial sweeteners- aspartame, acesulfame-K and binary sweetener blend of aspartame x acesulfame-K during storage. Acetoacetamide was also used in analysis of multiple sweeteners and their degradation products in lassi by HPLC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Simple Method For The Preparation Of Acetoacetate.
Lopez-Soriano FJ and Argiles JM.
Analytical Letters, 18(5), 589-592 (1985)
Assessment of stability of binary sweetener blend (aspartame x acesulfame-K) during storage in whey lemon beverage.
Arora S, et al.
Journal of Food Science and Technology, 50(4), 1-7 (2011)
V George et al.
Journal of food science and technology, 47(4), 408-413 (2010-08-01)
A solid phase extraction method using C18 cartridges was standardized for the isolation of multiple sweeteners (aspartame, acesulfame-K and saccharin) and their degradation products (diketopiperazine, Lphenylalanine, acetoacetamide and 2-sulfobenzoic acid) from lassi. Analytical conditions for HPLC were standardized over C18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service