Skip to Content
Merck
All Photos(1)

Key Documents

328359

Sigma-Aldrich

Perfluorokerosene

low-boiling

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 101.00

CHF 101.00


Estimated to ship on22 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
CHF 101.00

About This Item

MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

CHF 101.00


Estimated to ship on22 April 2025


Request a Bulk Order

Quality Level

form

liquid

refractive index

n20/D 1.336 (lit.)

bp

70-240 °C

density

1.835 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Perfluorokerosene was used as:
  • internal reference for negative ion mass spectrometry[1]
  • mulling agent during the analysis of urinary calculi by difuse reflectance infrared Fourier transform spectroscopy[2]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perfluorokerosine. Useful internal reference for negative ion mass spectrometry.
Gohlke RS and Thompson LH.
Analytical Chemistry, 40(6), 1004-1005 (1968)
M Berthelot et al.
Clinical chemistry, 33(6), 780-783 (1987-06-01)
We investigated the application of diffuse reflectance infrared Fourier transform spectroscopy to analysis of urinary calculi and compared its operation with that of older sampling techniques (pellet, mull, and attenuated total reflectance). The new method requires shorter sample preparation time

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service