Skip to Content
Merck
All Photos(2)

Key Documents

325090

Sigma-Aldrich

Methyl (R)-(+)-3-bromo-2-methylpropionate

97%

Synonym(s):

(+)-Methyl (R)-β-bromoisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH(CH3)CO2CH3
CAS Number:
Molecular Weight:
181.03
Beilstein:
1720833
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]20/D +16°, neat

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.455 (lit.)

bp

85 °C/37 mmHg (lit.)

density

1.422 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

COC(=O)[C@@H](C)CBr

InChI

1S/C5H9BrO2/c1-4(3-6)5(7)8-2/h4H,3H2,1-2H3/t4-/m0/s1

InChI key

FKWNAVCXZSQYTA-BYPYZUCNSA-N

Application

Methyl (R)-(+)-3-bromo-2-methylpropionate can be used as a reactant to prepare:
  • (S)-3-(6-Methoxy-pyridin-3-yl)-2-methylpropionic acid methylester by reacting with 5-bromo-2-methoxy pyridine via nucleophilic displacement reaction.[1]
  • Decahydroisoquinoline derivative (NVP-ACQ090) as a potent antagonist of somatostatin sst3 receptor.[1]

It can also be used as a reference compound in the enantioselective alkylation reaction of hydrophobic vitamin B12 derivatives.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A 1H NMR and molecular modelling investigation of diastereotopic methylene hydrogen atoms.
Tormena CF, et al.
Magnetic Resonance in Chemistry, 40(4), 279-283 (2002)
The development of a practical synthesis of the potent and selective somatostatin sst3 receptor antagonist [4-(3, 4-difluoro-phenyl)-piperazine-1-yl]-
Banziger M, et al.
Tetrahedron Asymmetry, 14(22), 3469-3477 (2003)
Hydrophobic vitamin B12. Part 12. Preparation, characterization and enantioselective alkylation of strapped hydrophobic-vitamin B12.
Murakami Y, et al.
J. Chem. Soc. Perkin Trans. II, 6, 1175-1183 (1995)
Hydrophobic vitamin B12. Part 12. Preparation, characterization and enantioselective alkylation of strapped hydrophobic-vitamin B12
Murakami Y, et al.
J. Chem. Soc. Perkin Trans. II, 14(6), 1175-1183 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service