Recommended Products
form
powder
concentration
38.0-44.0% Ca (complexometric)
InChI
1S/FH.K/h1H;/q;+1/p-1
InChI key
NROKBHXJSPEDAR-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Promotes efficient and mild fluorination of a variety of organic bromides and chlorides.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of the Chemical Society. Chemical Communications, 791-791 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service