Skip to Content
Merck
All Photos(1)

Key Documents

303097

Sigma-Aldrich

4,5-Dichloro-3-hydroxypyridazine

98%

Synonym(s):

4,5-Dichloro-3(2H)-pyridazinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2Cl2N2O
CAS Number:
Molecular Weight:
164.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

204-206 °C (lit.)

functional group

chloro

SMILES string

ClC1=C(Cl)C(=O)NN=C1

InChI

1S/C4H2Cl2N2O/c5-2-1-7-8-4(9)3(2)6/h1H,(H,8,9)

InChI key

VJWXIRQLLGYIDI-UHFFFAOYSA-N

General description

The solid phase FT-IR and FT-Raman spectra of 4,5-dichloro-3-hydroxypyridazine was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Krishnakumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(11-12), 2526-2532 (2005-07-27)
The solid phase FT-IR and FT-Raman spectra of 4,5-dichloro-3-hydroxypyridazine have been recorded in the regions 4000-400 cm(-1) and 3500-100 cm(-1), respectively. The spectra were interpreted with the aid of normal coordinate analysis following a full structure optimization and force field

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service