Skip to Content
Merck
All Photos(1)

Key Documents

30192

Sigma-Aldrich

(R)-2-Aminohexane

≥97.0% (GC)

Synonym(s):

(R)-2-Hexylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15N
CAS Number:
Molecular Weight:
101.19
Beilstein:
5237601
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥97.0% (GC)

optical purity

enantiomeric ratio: >99.0:1.0 (GC)

SMILES string

CCCC[C@@H](C)N

InChI

1S/C6H15N/c1-3-4-5-6(2)7/h6H,3-5,7H2,1-2H3/t6-/m1/s1

InChI key

WGBBUURBHXLGFM-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F

Flash Point(C)

54 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service