Skip to Content
Merck
All Photos(1)

Documents

288179

Sigma-Aldrich

Chromium(III) chloride tetrahydrofuran complex (1:3)

97%

Synonym(s):

Tetrahydrofuran- trichlorochromium (3:1), oxolane trichlorochromium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24Cl3CrO3
CAS Number:
Molecular Weight:
374.67
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

143 °C (dec.) (lit.)

SMILES string

Cl[Cr](Cl)Cl.C1CCOC1.C2CCOC2.C3CCOC3

InChI

1S/3C4H8O.3ClH.Cr/c3*1-2-4-5-3-1;;;;/h3*1-4H2;3*1H;/q;;;;;;+3/p-3

InChI key

CYOMBOLDXZUMBU-UHFFFAOYSA-K

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service