287091
Pyridoxamine dihydrochloride
Synonym(s):
4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H12N2O2 · 2HCl
CAS Number:
Molecular Weight:
241.11
Beilstein:
3632748
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
Recommended Products
impurities
may contain ≤10% water
mp
224-226 °C (dec.) (lit.)
SMILES string
Cl[H].Cl[H].Cc1ncc(CO)c(CN)c1O
InChI
1S/C8H12N2O2.2ClH/c1-5-8(12)7(2-9)6(4-11)3-10-5;;/h3,11-12H,2,4,9H2,1H3;2*1H
InChI key
HNWCOANXZNKMLR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service