Skip to Content
Merck
All Photos(1)

Key Documents

284475

Sigma-Aldrich

(2R,4R,5S)-(+)-2-Chloro-3,4-dimethyl-5-phenyl-1,3,2-oxazaphospholidine 2-sulfide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClNOPS
CAS Number:
Molecular Weight:
261.71
UNSPSC Code:
12352103

Assay

98%

optical activity

[α]20/D +120°, c = 1 in chloroform

mp

127-129 °C (lit.)

SMILES string

C[C@@H]1[C@@H](O[P@@](Cl)(=S)N1C)c2ccccc2

Looking for similar products? Visit Product Comparison Guide

Application

Derivatizing reagent for the 31P NMR determination of enantiomeric purities of alcohols and amines.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 106, 5019-5019 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service