Skip to Content
Merck
All Photos(2)

Key Documents

277053

Sigma-Aldrich

(S)-(+)-2-(Methoxymethyl)pyrrolidine

99%

Synonym(s):

O-Methyl-L-prolinol, SMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
Beilstein:
3587324
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]20/D +2.4°, c = 2 in benzene

refractive index

n20/D 1.4457 (lit.)

bp

62 °C/40 mmHg (lit.)

density

0.933 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

COC[C@@H]1CCCN1

InChI

1S/C6H13NO/c1-8-5-6-3-2-4-7-6/h6-7H,2-5H2,1H3/t6-/m0/s1

InChI key

CHPRFKYDQRKRRK-LURJTMIESA-N

Application

(S)-(+)-2-(Methoxymethyl)pyrrolidine may be used to prepare planar chiral 2-phospha[3]ferrocenophanes.[1] It may also be used as an organocatalyst in the cross-aldol reaction of dihydroxyacetone with p-nitrobenzaldehyde to form the corresponding polyol.[2] SMP reacts with benzoylformic acid to form (S)-N-benzoylformyl-2-(methoxymethyl)-pyrrolidine.[3]
Chiral auxiliary for asymmetric synthesis of propargyl alcohols[4] and 4-nitrocyclohexanones,[5] and for asymmetric Michael additions to enones.[6]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Kawauchi et al.
Oncogene, 36(37), 5231-5242 (2017-05-16)
Group3 medulloblastoma (MB
Kirsten E Bell et al.
PloS one, 12(7), e0181387-e0181387 (2017-07-19)
Protein and other compounds can exert anabolic effects on skeletal muscle, particularly in conjunction with exercise. The objective of this study was to evaluate the efficacy of twice daily consumption of a protein-based, multi-ingredient nutritional supplement to increase strength and
Synthesis, 1242-1242 (1992)
Roberta Fraioli et al.
Scientific reports, 7(1), 16363-16363 (2017-11-29)
Engineering the interface between biomaterials and tissues is important to increase implant lifetime and avoid failures and revision surgeries. Permanent devices should enhance attachment and differentiation of stem cells, responsible for injured tissue repair, and simultaneously discourage bacterial colonization; this
Cian Vyas et al.
Materials science & engineering. C, Materials for biological applications, 118, 111433-111433 (2020-12-02)
Polycaprolactone (PCL) scaffolds have been widely investigated for tissue engineering applications, however, they exhibit poor cell adhesion and mechanical properties. Subsequently, PCL composites have been produced to improve the material properties. This study utilises a natural material, Bombyx mori silk

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service