Recommended Products
form
powder
Quality Level
solubility
methanol: 1 mg/mL
λmax
611 nm
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
Nc1cc2OC3=CC(=O)C(=CC3=Nc2cc1-c4ccc(O)cc4O)c5ccc(O)cc5O
InChI
1S/C24H16N2O6/c25-17-9-23-18(7-15(17)13-3-1-11(27)5-20(13)29)26-19-8-16(22(31)10-24(19)32-23)14-4-2-12(28)6-21(14)30/h1-10,27-30H,25H2
InChI key
ABNVFGQXCGFSFH-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service