Skip to Content
Merck
All Photos(1)

Key Documents

274607

Sigma-Aldrich

Bis(4-methoxyphenyl)phosphinic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OC6H4)2P(O)OH
CAS Number:
Molecular Weight:
278.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Assay

99%

mp

185-187 °C (lit.)

SMILES string

COc1ccc(cc1)P(O)(=O)c2ccc(OC)cc2

InChI

1S/C14H15O4P/c1-17-11-3-7-13(8-4-11)19(15,16)14-9-5-12(18-2)6-10-14/h3-10H,1-2H3,(H,15,16)

InChI key

BFPBWJGVRNQWEK-UHFFFAOYSA-N

Application

Bis(4-methoxyphenyl)phosphinic acid was used to treat the chemical surface of porphyrin-sensitised titania films after dye adsorption to enhance the efficiency of dye-sensitized solar cells (DSSC)[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved performance of porphyrin-based dye sensitised solar cells by phosphinic acid surface treatment.
Energy & Environmental Science, 2(10), 1069-1073 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service