Skip to Content
Merck
All Photos(1)

Key Documents

272965

Sigma-Aldrich

2,4-Dimethyl-2,3-pentadiene

97%

Synonym(s):

Tetramethylallene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=C=C(CH3)2
CAS Number:
Molecular Weight:
96.17
Beilstein:
1733258
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.440 (lit.)

bp

87-88 °C (lit.)

density

0.701 g/mL at 25 °C (lit.)

SMILES string

CC(C)=C=C(C)C

InChI

1S/C7H12/c1-6(2)5-7(3)4/h1-4H3

InChI key

DZSNJASVIURWOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dimethyl-2,3-pentadiene was used in the synthesis of 3-(2,4-dimethyl-1,2-pentadienyl) and 2-(3-methyl-1,2-butadienyl) sulfinic acids via ene addition reaction in liquid sulphur dioxide[1].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon P Neville et al.
The Journal of chemical physics, 144(1), 014305-014305 (2016-01-10)
We report a joint experimental and theoretical study on the ultrafast excited state dynamics of allene and a series of its methylated analogues (1,2-butadiene, 1,1-dimethylallene, and tetramethylallene) in order to elucidate the conical intersection mediated dynamics that give rise to
Capozzi G, et al.
Tetrahedron Letters, 21(34), 3289-3292 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service