Skip to Content
Merck
All Photos(1)

Key Documents

272337

Sigma-Aldrich

Spiro[2.4]hepta-4,6-diene

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8
CAS Number:
Molecular Weight:
92.14
Beilstein:
969332
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

technical grade

Assay

90%

form

solid

density

0.92 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CC12C=CC=C2

InChI

1S/C7H8/c1-2-4-7(3-1)5-6-7/h1-4H,5-6H2

InChI key

KPDOKSJMJMDRQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service