264148
Triphenylborane–Sodium hydroxide adduct solution
6-10 wt. % in H2O
About This Item
composition
NaCl, 6-7%
NaOH, 3-4%
concentration
6-10 wt. % in H2O
refractive index
n20/D 1.378
density
1.07 g/mL at 25 °C
SMILES string
[OH-].[Na+].c1ccc(cc1)B(c2ccccc2)c3ccccc3
InChI
1S/C18H15B.Na.H2O/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h1-15H;;1H2/q;+1;/p-1
InChI key
NBZUMVYYQTYYAN-UHFFFAOYSA-M
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service