Skip to Content
Merck
All Photos(2)

Key Documents

26230

Sigma-Aldrich

Chloro(3-chloropropyl)dimethylsilane

technical, ≥95% (GC)

Synonym(s):

(3-Chloropropyl)dimethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12Cl2Si
CAS Number:
Molecular Weight:
171.14
Beilstein:
506022
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥95% (GC)

form

liquid

refractive index

n20/D 1.450

bp

177-179 °C (lit.)

density

1.046 g/mL at 20 °C (lit.)

functional group

chloro

SMILES string

C[Si](C)(Cl)CCCCl

InChI

1S/C5H12Cl2Si/c1-8(2,7)5-3-4-6/h3-5H2,1-2H3

InChI key

BJLJNLUARMMMLW-UHFFFAOYSA-N

Application

Chloro(3-chloropropyl)dimethylsilane can be used as:
  • A starting material to prepare misonidazole-based 18F-radiolabeled organosilicon compounds as positron emission tomography (PET) ligands for medical imaging applications.[1]
  • An intermediate in the synthesis of block copolymers.[2][3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Fauquignon et al.
Polymers, 11(12) (2019-12-11)
In the emerging field of hybrid polymer/lipid vesicles, relatively few copolymers have been evaluated regarding their ability to form these structures and the resulting membrane properties have been scarcely studied. Here, we present the synthesis and self-assembly in solution of
Design of silicon-based misonidazole analogues and 18F-radiolabelling
Bohn P, et al.
Nuclear Medicine and Biology, 36(8), 895-905 (2009)
Universal methodology for block copolymer synthesis
Bellas V and Rehahn M
Macromolecular Rapid Communications, 28(13), 1415-1421 (2007)
Large and Giant Unilamellar Vesicle (s) Obtained by Self-Assembly of Poly (dimethylsiloxane)-b-poly (ethylene oxide) Diblock Copolymers, Membrane Properties and Preliminary Investigation of Their Ability to Form Hybrid Polymer/Lipid Vesicles
Fauquignon M, et al.
Polymers, 11(12), 2013-2013 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service