Skip to Content
Merck
All Photos(1)

Key Documents

255866

Sigma-Aldrich

2-(1-Cyclohexenyl)ethylamine

97%

Synonym(s):

1-Cyclohexene-1-ethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9CH2CH2NH2
CAS Number:
Molecular Weight:
125.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4865 (lit.)

bp

53-54 °C/2.5 mmHg (lit.)

density

0.898 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCCC1=CCCCC1

InChI

1S/C8H15N/c9-7-6-8-4-2-1-3-5-8/h4H,1-3,5-7,9H2

InChI key

IUDMXOOVKMKODN-UHFFFAOYSA-N

Application

2-(1-Cyclohexenyl)ethylamine has been employed:
  • as substrate for allylic hydroxylation reaction[1]
  • in preparation of thin films and single crystals of 2-(1-cyclohexenyl)ethyl ammonium lead iodide, used to fabricate optoelectronic-compatible heterostructures[2]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Pradeesh et al.
Optics express, 17(24), 22171-22178 (2009-12-10)
Optoelectronic-compatible heterostructures are fabricated from layered inorganic-organic multiple quantum wells (IO-MQW) of Cyclohexenyl ethyl ammonium lead iodide, (C(6)H(9)C(2)H(4)NH(3))(2)PbI(4) (CHPI). These hybrids possess strongly-resonant optical features, are thermally stable and compatible with hybrid photonics assembly. Room-temperature strong-coupling is observed when these
Facile stereoselective allylic hydroxylation by dopamine. beta.-monooxygenase.
Sirimanne SR and May SW.
Journal of the American Chemical Society, 110(22), 7560-7561 (1988)
S R Sirimanne et al.
The Biochemical journal, 306 ( Pt 1), 77-85 (1995-02-15)
The reaction of dopamine beta-monooxygenase (DBM; EC 1.14.17.1) with the prototypical non-conjugated olefinic substrate, 2-(1-cyclohexenyl)ethylamine (CyHEA) [see Sirimanne and May (1988) J. Am. Chem. Soc. 110, 7560-7561], was characterized. CyHEA undergoes facile DBM-catalysed allylic hydroxylation to form (R)-2-amino-1-(1-cyclohexenyl)ethanol (CyHEA-OH) without

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service