Skip to Content
Merck
All Photos(1)

Key Documents

249483

Sigma-Aldrich

2-(Trifluoromethyl)benzoyl chloride

99%

Synonym(s):

α,α,α-Trifluoro-o-toluoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4COCl
CAS Number:
Molecular Weight:
208.56
Beilstein:
881040
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.479 (lit.)

bp

84-85 °C/16 mmHg (lit.)

density

1.416 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccccc1C(Cl)=O

InChI

1S/C8H4ClF3O/c9-7(13)5-3-1-2-4-6(5)8(10,11)12/h1-4H

InChI key

MXIUWSYTQJLIKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(Trifluoromethyl)benzoyl chloride may be used in chemical synthesis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Premnath et al.
Interdisciplinary sciences, computational life sciences, 9(1), 130-139 (2016-01-05)
The commenced work deals with the synthesis, characterization and evaluation of biological activities of 4-amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one. The synthesis was done by the condensation of aromatic acid chlorides with 4-aminoantipyrine. The structures of synthesized derivatives were elucidated using IR, Mass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service