Skip to Content
Merck
All Photos(1)

Key Documents

247022

Sigma-Aldrich

Boron trichloride methyl sulfide complex solution

2.0 M in methylene chloride

Synonym(s):

Dimethyl sulfide-trichloroborane, Trichloroborane-methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S·BCl3
CAS Number:
Molecular Weight:
179.30
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

concentration

2.0 M in methylene chloride

density

1.344 g/mL at 25 °C

SMILES string

C[S+](C)[B-](Cl)(Cl)Cl

InChI

1S/C2H6BCl3S/c1-7(2)3(4,5)6/h1-2H3

InChI key

YNDYUDLLOGOQNW-UHFFFAOYSA-N

Application

Convenient reagent for dealkylation of aryl ethers.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tetrahedron Letters, 21, 3731-3731 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service