Skip to Content
Merck
All Photos(1)

Key Documents

245437

Sigma-Aldrich

α-(Cyanoimino)-3,4-dichlorophenethylamine

99%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CHF 269.00

CHF 269.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
CHF 269.00

About This Item

Linear Formula:
Cl2C6H3CH2C(=NCN)NH2
CAS Number:
Molecular Weight:
228.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 269.00


Estimated to ship on01 May 2025


Request a Bulk Order

Quality Level

Assay

99%

form

solid

mp

143-145 °C (lit.)

SMILES string

N\C(Cc1ccc(Cl)c(Cl)c1)=N/C#N

InChI

1S/C9H7Cl2N3/c10-7-2-1-6(3-8(7)11)4-9(13)14-5-12/h1-3H,4H2,(H2,13,14)

InChI key

FPJZOLMBOBRFFS-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service