Skip to Content
Merck
All Photos(1)

Key Documents

24377

Sigma-Aldrich

2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate

≥95.0% (AT)

Synonym(s):

CIB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10BClF4N2
CAS Number:
Molecular Weight:
220.40
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (AT)

form

solid

reaction suitability

reaction type: Coupling Reactions

mp

175-177 °C (lit.)
175-178 °C

solubility

acetonitrile: 0.1 g/mL, clear

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.CN1CC[N+](C)=C1Cl

InChI

1S/C5H10ClN2.BF4/c1-7-3-4-8(2)5(7)6;2-1(3,4)5/h3-4H2,1-2H3;/q+1;-1

InChI key

UPLXKEIRISBKRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for:
Esterification and peptide coupling of sterically hindered amino acids
Preparation of efficient diazo-transfer reagents
Oxidative insertion for palladium and nickel catalyst synthesis
Preparation of catalysts for use in coupling reactions

Other Notes

Reagent for esterification of C-terminal amino acids to Wang resin without racemization and for coupling α,α-dimethyl amino acids[1][2]; Preparation of the benzotriazol-1-yloxy derivative, a new coupling reagent[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 33, 3177-3177 (1992)
K. Akaji et al.
Proc. 22nd Eur. Pept. Symp.: Peptides 1992, 220-220 (1992)
Y. Kiso et al.
Chemical & Pharmaceutical Bulletin, 38, 270-270 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service